accessibility menu, dialog, popup

UserName

H3CN(R)O(R)OH(R)OH(S)ONO

CAS RN 22423-26-3

IUPAC Name: (2R,4R,5R,6S)-5-hydroxy-4-(hydroxymethyl)-11-methyl-3,7-dioxa-1,9-diazatricyclo[6.4.0.0²,⁶]dodeca-8,11-dien-10-one
Synonyms: (2R,4R,5R,6S)-5-hydroxy-4-(hydroxymethyl)-11-methyl-3,7-dioxa-1,9-diazatricyclo[6.4.0.0²,⁶]dodeca-8,11-dien-10-one
Molecular Weight (g/mol): 240.22
Molecular Formula: C10H12N2O5
InChi Key: WLLOAUCNUMYOQI-JAGXHNFQSA-N
SMILES: CC1=CN2[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3OC2=NC1=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results