missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Nitroethoxy)tetrahydropyran 85.0+%, TCI America™

Catalog No. p-7133888
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
N06025G 5 g
N060225G 25 g
2 options

Catalog No. N06025G

Supplier: TCI America N06025G

Only null left

Chemical Identifiers

CAS 75233-61-3
Molecular Formula C7H13NO4
Molecular Weight (g/mol) 175.184
MDL Number MFCD00010858
InChI Key KQDXRRAVDLIMGM-UHFFFAOYSA-N
Synonym Tetrahydro-2-(2-nitroethoxy)-2H-pyran
PubChem CID 3748656
IUPAC Name 2-(2-nitroethoxy)oxane
SMILES C1CCOC(C1)OCC[N+](=O)[O-]

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 175.18
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Nitroethoxy)tetrahydropyran
TSCA No
Recommended Storage Freezer