missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Naphthyl)indole, 98%

Catalog No. AAB2043406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2043406 5 g
AAB2043403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2043406 Supplier Thermo Scientific Chemicals Supplier No. B2043406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23746-81-8 | C18H13N | 243.31 g/mol

It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

Solubility
Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 23746-81-8
Molecular Formula C18H13N
Molecular Weight (g/mol) 243.31
MDL Number MFCD00124144
InChI Key CACDYUNTCMPDMI-UHFFFAOYSA-N
Synonym 2-2-naphthyl indole, 2-2-naphthyl-1h-indole, 2-naphthalen-2-yl-1h-indole, acmc-1chvp, 2-2-naphthyl-indol, maybridge1_005904, 2-2-indolyl naphthalene, 2-1h-indol-2-yl naphthalene, 2-2-naphthalenyl-1h-indole
PubChem CID 265232
SMILES N1C2=CC=CC=C2C=C1C1=CC=C2C=CC=CC2=C1

Specifications

Quantity 5 g
Solubility Information Soluble in DMSO.
IUPAC Name 2-(naphthalen-2-yl)-1H-indole
Formula Weight 243.31
Percent Purity 98%
Chemical Name or Material 2-(2-Naphthyl)indole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only