accessibility menu, dialog, popup

UserName

NH

CAS RN 23746-81-8

IUPAC Name: 2-(naphthalen-2-yl)-1H-indole
Synonyms: 2-(naphthalen-2-yl)-1H-indole
Molecular Weight (g/mol): 243.31
Molecular Formula: C18H13N
InChi Key: CACDYUNTCMPDMI-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2C=C1C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)