missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Naphthoxy)ethanol, 98+%

CAS: 93-20-9 | C12H12O2 | 188.226 g/mol

Supplier:  Thermo Scientific Chemicals A1163614

 View more versions of this product

Catalog No. AAA1163614


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2- (2-Naphthoxy)ethanol
72°C to 76°C
MFCD00016809
2086973
2-2-naphthyloxy ethanol, 2-2-naphthoxy ethanol, 2-naphthalen-2-yloxy ethanol, anavenol, 2-2-hydroxyethoxy naphthalene, ethanol, 2-2-naphthalenyloxy, 2-2-naphthalenyloxy ethanol, unii-7m7cut7ccu, ethylene glycol mono-2-naphthyl ether, .beta.-naphthoxyethanol
C1=CC=C2C=C(C=CC2=C1)OCCO
188.226
188.23
93-20-9
C12H12O2
25 g
14,6413
BQPBZDSDFCDSAO-UHFFFAOYSA-N
2-naphthalen-2-yloxyethanol
7131
≥98%

Chemical Identifiers

93-20-9
188.226
BQPBZDSDFCDSAO-UHFFFAOYSA-N
7131
C1=CC=C2C=C(C=CC2=C1)OCCO
C12H12O2
MFCD00016809
2-2-naphthyloxy ethanol, 2-2-naphthoxy ethanol, 2-naphthalen-2-yloxy ethanol, anavenol, 2-2-hydroxyethoxy naphthalene, ethanol, 2-2-naphthalenyloxy, 2-2-naphthalenyloxy ethanol, unii-7m7cut7ccu, ethylene glycol mono-2-naphthyl ether, .beta.-naphthoxyethanol
2-naphthalen-2-yloxyethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 202-228-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only