missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Naphthoxy)ethanol, 98+%

Catalog No. p-7081553
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1163606 5 g
AAA1163614 25 g
AAA1163622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1163606 Supplier Thermo Scientific Chemicals Supplier No. A1163606

CAS: 93-20-9 | C12H12O2 | 188.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93-20-9
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD00016809
InChI Key BQPBZDSDFCDSAO-UHFFFAOYSA-N
Synonym 2-2-naphthyloxy ethanol, 2-2-naphthoxy ethanol, 2-naphthalen-2-yloxy ethanol, anavenol, 2-2-hydroxyethoxy naphthalene, ethanol, 2-2-naphthalenyloxy, 2-2-naphthalenyloxy ethanol, unii-7m7cut7ccu, ethylene glycol mono-2-naphthyl ether, .beta.-naphthoxyethanol
PubChem CID 7131
IUPAC Name 2-naphthalen-2-yloxyethanol
SMILES C1=CC=C2C=C(C=CC2=C1)OCCO

Specifications

Melting Point 72°C to 76°C
Quantity 5 g
Beilstein 2086973
Merck Index 14,6413
Formula Weight 188.23
Percent Purity ≥98%
Chemical Name or Material 2- (2-Naphthoxy)ethanol
EINECSNumber 202-228-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only