missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-(Methylimino)bis(N,N-di-n-octylacetamide) 95.0+%, TCI America™
Supplier: TCI America M24765ML
Specifications
| 2,2′-(Methylimino)bis(N,N-di-n-octylacetamide) | |
| C37H75N3O2 | |
| 5 mL | |
| VYQLYCLTFJDXGV-UHFFFAOYSA-N | |
| 2-{[(dioctylcarbamoyl)methyl](methyl)amino}-N,N-dioctylacetamide | |
| 71551252 | |
| ≥95.0% (GC,T) |
| 1000668-90-5 | |
| MFCD28386110 | |
| MIDOA | |
| CCCCCCCCN(CCCCCCCC)C(=O)CN(C)CC(=O)N(CCCCCCCC)CCCCCCCC | |
| 594.03 | |
| 594.03 | |
| Liquid |
Chemical Identifiers
| 1000668-90-5 | |
| 594.03 | |
| VYQLYCLTFJDXGV-UHFFFAOYSA-N | |
| 71551252 | |
| CCCCCCCCN(CCCCCCCC)C(=O)CN(C)CC(=O)N(CCCCCCCC)CCCCCCCC |
| C37H75N3O2 | |
| MFCD28386110 | |
| MIDOA | |
| 2-{[(dioctylcarbamoyl)methyl](methyl)amino}-N,N-dioctylacetamide |
Safety and Handling
TSCA : No