accessibility menu, dialog, popup

UserName

CAS RN 1000668-90-5

IUPAC Name: 2-{[(dioctylcarbamoyl)methyl](methyl)amino}-N,N-dioctylacetamide
Synonyms: 2-{[(dioctylcarbamoyl)methyl](methyl)amino}-N,N-dioctylacetamide
Molecular Weight (g/mol): 594.03
Molecular Formula: C37H75N3O2
InChi Key: VYQLYCLTFJDXGV-UHFFFAOYSA-N
SMILES: CCCCCCCCN(CCCCCCCC)C(=O)CN(C)CC(=O)N(CCCCCCCC)CCCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results