missing translation for 'onlineSavingsMsg'
Learn More

2,2'-(Methylimino)bis(N,N-di-n-octylacetamide) 95.0+%, TCI America™

Catalog No. M24761ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24761ML 1 mL
M24765ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24761ML Supplier TCI America Supplier No. M24761ML
Only null left

Chemical Identifiers

CAS 1000668-90-5
Molecular Formula C37H75N3O2
Molecular Weight (g/mol) 594.03
MDL Number MFCD28386110
InChI Key VYQLYCLTFJDXGV-UHFFFAOYSA-N
Synonym MIDOA
PubChem CID 71551252
IUPAC Name 2-{[(dioctylcarbamoyl)methyl](methyl)amino}-N,N-dioctylacetamide
SMILES CCCCCCCCN(CCCCCCCC)C(=O)CN(C)CC(=O)N(CCCCCCCC)CCCCCCCC

Specifications

Quantity 1 mL
Formula Weight 594.03
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2′-(Methylimino)bis(N,N-di-n-octylacetamide)