missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Methylenebis[6-(1-methylcyclohexyl)-p-cresol] 95.0+%, TCI America™

Catalog No. M130125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M130125G 25 g
M1301500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M130125G Supplier TCI America Supplier No. M130125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-62-3
Molecular Formula C29H40O2
Molecular Weight (g/mol) 420.64
MDL Number MFCD00151797
InChI Key PHXLONCQBNATSL-UHFFFAOYSA-N
Synonym 2,2'-methylenebis 6-1-methylcyclohexyl-p-cresol, ionox wsp, nonox wsp, bisalkofen mtsp, unii-7tir91vdi9, 7tir91vdi9, p-cresol, 2,2'-methylenebis 6-1-methylcyclohexyl, 2,2'-methylenebis 4-methyl-6-1-methylcyclohexyl phenol
PubChem CID 6486
IUPAC Name 2-{[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methyl}-4-methyl-6-(1-methylcyclohexyl)phenol
SMILES CC1=CC(=C(O)C(CC2=C(O)C(=CC(C)=C2)C2(C)CCCCC2)=C1)C1(C)CCCCC1

Specifications

Melting Point 138°C
Quantity 25 g
Formula Weight 420.64
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Methylenebis[6-(1-methylcyclohexyl)-p-cresol]
EINECSNumber (4)-0888
RTECSNumber SM0400000
TSCA Yes