missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-5-nitroanilino)-4-(3-pyridyl)pyrimidine 98.0+%, TCI America™

Catalog No. M25845G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25845G 5 g
M25841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25845G Supplier TCI America Supplier No. M25845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 152460-09-8
Molecular Formula C16H13N5O2
Molecular Weight (g/mol) 307.313
MDL Number MFCD02179269
InChI Key OJITWRFPRCHSMX-UHFFFAOYSA-N
Synonym n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimidin-2-amine, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidine-amine, chembl20426, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidineamine, n-2-methyl-5-nitrophenyl-4-pyridin-3-ylpyrimidin-2-amine, 2-methyl-4-nitro-phenyl-4-pyridin-3-yl-pyrimidin-2-yl-amine, 2-methyl-5-nitro-n-4-pyridin-3-yl pyrimidin-2-yl benzenamine, 2-pyrimidinamine, n-2-methyl-5-nitrophenyl-4-3-pyridinyl, n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimid, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidinamine
PubChem CID 10686149
IUPAC Name N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])NC2=NC=CC(=N2)C3=CN=CC=C3

Specifications

Melting Point 194°C
Color Yellow
Quantity 5 g
Formula Weight 307.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Methyl-5-nitroanilino)-4-(3-pyridyl)pyrimidine