accessibility menu, dialog, popup

UserName

CH3NOONHNNN

CAS RN 152460-09-8

IUPAC Name: N-(2-methyl-5-nitrophenyl)-4-(pyridin-3-yl)pyrimidin-2-amine
Synonyms: N-(2-methyl-5-nitrophenyl)-4-(pyridin-3-yl)pyrimidin-2-amine
Molecular Weight (g/mol): 307.31
Molecular Formula: C16H13N5O2
InChi Key: OJITWRFPRCHSMX-UHFFFAOYSA-N
SMILES: CC1=C(NC2=NC=CC(=N2)C2=CN=CC=C2)C=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results