missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyphenoxy)ethylamine 95.0+%, TCI America™

Catalog No. M20045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20045G 5 g
M200425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20045G Supplier TCI America Supplier No. M20045G

Chemical Identifiers

CAS 1836-62-0
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00235185
InChI Key CKJRKLKVCHMWLV-UHFFFAOYSA-N
Synonym 2-2-methoxyphenoxy ethylamine, 2-2-methoxyphenoxy ethanamine, 1-2-aminoethoxy-2-methoxybenzene, 2-2-methoxyphenoxy ethyl amine, 2-2-methoxy-phenoxy-ethylamine, ethanamine, 2-2-methoxyphenoxy, unii-20s0vx4qt9, 2-2'-methoxy phenoxyethylamine, 2-2-methoxyphenoxy ethan-1-amine
PubChem CID 1713005
IUPAC Name 2-(2-methoxyphenoxy)ethanamine
SMILES COC1=CC=CC=C1OCCN

Specifications

Boiling Point 98°C
Quantity 5 g
Formula Weight 167.21
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Methoxyphenoxy)ethylamine