missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Methoxyphenoxy)ethylamine Hydrochloride Hydrate 98.0+%, TCI America™
$44.73 - $132.35
Chemical Identifiers
| CAS | 64464-07-9 |
|---|---|
| Molecular Formula | C9H14ClNO2 |
| Molecular Weight (g/mol) | 203.67 |
| MDL Number | MFCD01631120 |
| InChI Key | KNWPXZOMSZABHD-UHFFFAOYSA-N |
| Synonym | 2-(2-Aminoethoxy)anisole Hydrochloride, 1-(2-Aminoethoxy)-2-methoxybenzene Hydrochloride |
| PubChem CID | 2762734 |
| IUPAC Name | 2-(2-methoxyphenoxy)ethan-1-amine hydrochloride |
| SMILES | Cl.COC1=CC=CC=C1OCCN |
Chemical Identifiers
| 64464-07-9 | |
| 203.67 | |
| KNWPXZOMSZABHD-UHFFFAOYSA-N | |
| 2762734 | |
| Cl.COC1=CC=CC=C1OCCN |
| C9H14ClNO2 | |
| MFCD01631120 | |
| 2-(2-Aminoethoxy)anisole Hydrochloride, 1-(2-Aminoethoxy)-2-methoxybenzene Hydrochloride | |
| 2-(2-methoxyphenoxy)ethan-1-amine hydrochloride |
Specifications
| 64464-07-9 | |
| C9H14ClNO2 | |
| 5 g | |
| KNWPXZOMSZABHD-UHFFFAOYSA-N | |
| 2-(2-methoxyphenoxy)ethan-1-amine hydrochloride | |
| 2762734 | |
| ≥98.0% (T) | |
| 2-(2-Methoxyphenoxy)ethylamine Hydrochloride Hydrate |
| White | |
| MFCD01631120 | |
| 2-(2-Aminoethoxy)anisole Hydrochloride, 1-(2-Aminoethoxy)-2-methoxybenzene Hydrochloride | |
| Cl.COC1=CC=CC=C1OCCN | |
| 203.67 | |
| 203.67 | |
| Crystalline Powder |
Safety and Handling
TSCA : No