accessibility menu, dialog, popup

UserName

CH3OOH2NHCl

CAS RN 64464-07-9

IUPAC Name: 2-(2-methoxyphenoxy)ethan-1-amine hydrochloride
Synonyms: 2-(2-methoxyphenoxy)ethanamine hydrochloride
Molecular Weight (g/mol): 203.67
Molecular Formula: C9H14ClNO2
InChi Key: KNWPXZOMSZABHD-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=CC=C1OCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)