missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Methoxyphenoxy)ethylamine Hydrochloride Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 203.67 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(2-Methoxyphenoxy)ethylamine Hydrochloride Hydrate |
Safety and Handling
| TSCA | No |