missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyethoxy)ethanol, 99%

Catalog No. p-4510142
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149370010 1 L Glass Bottle
AC149370025 2.5 L Glass Bottle
AC149370100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC149370010 Supplier Thermo Scientific Chemicals Supplier No. 149370010

CAS: 111-77-3 | C5H12O3 | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-77-3
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00002871,MFCD00084416
InChI Key SBASXUCJHJRPEV-UHFFFAOYSA-N
Synonym 2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm
PubChem CID 8134
ChEBI CHEBI:44836
IUPAC Name 2-(2-methoxyethoxy)ethanol
SMILES COCCOCCO

Specifications

Melting Point -70.0°C
Density 1.0100g/mL
Boiling Point 194.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4250 to 1.4270
Linear Formula CH3OCH2CH2OCH2CH2OH
Quantity 1 L
Beilstein 01,IV,2392
Specific Gravity 1.01
Merck Index 15,3136
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,glycerol,ether,acetone,and dmf
Viscosity 3.9 mPa.s (20°C)
Formula Weight 120.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(2-Methoxyethoxy)ethanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 203-906-6

RUO – Research Use Only