missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyethoxy)ethanol, 98%, Spectrum™ Chemical

Catalog No. p-7165858
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
20 L
4 L
500 mL
Packaging:
Amber Glass Bottle
Steel Can
Catalog No. Quantity Packaging
18608150 500 mL Amber Glass Bottle
18608149 4 L Amber Glass Bottle
18608151 20 L Steel Can
3 options

Catalog No. 18608150

Supplier: Spectrum Chemical Mfg Cor M1222500MLGL

Only null left

Chemical Identifiers

CAS 111-77-3
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
InChI Key SBASXUCJHJRPEV-UHFFFAOYSA-N
IUPAC Name 2-(2-methoxyethoxy)ethan-1-ol
SMILES COCCOCCO

Specifications

Acidity 0.0001
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.