missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™

Catalog No. I06531G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06531G Supplier TCI America Supplier No. I06531G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 857934-82-8
Molecular Formula C12H16BIO2
Molecular Weight (g/mol) 329.97
MDL Number MFCD09038529
InChI Key HSQZIRZPACYGJJ-UHFFFAOYSA-N
Synonym 1-Iodo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-Iodophenylboronic Acid Pinacol Ester
PubChem CID 12158590
IUPAC Name 2-(2-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1I

Specifications

Color Yellow
Quantity 1 g
Formula Weight 329.97
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material 2-(2-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane