missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Hydroxyethyl)-1-methylpyrrolidine 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 111°C |
| Quantity | 10 mL |
| Formula Weight | 129.20 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-(2-Hydroxyethyl)-1-methylpyrrolidine |
Safety and Handling
| TSCA | No |