missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole 98.0+%, TCI America™
$65.24 - $451.10
Chemical Identifiers
| CAS | 3147-75-9 |
|---|---|
| Molecular Formula | C20H25N3O |
| Molecular Weight (g/mol) | 323.44 |
| MDL Number | MFCD00013338 |
| InChI Key | IYAZLDLPUNDVAG-UHFFFAOYSA-N |
| PubChem CID | 62485 |
| IUPAC Name | 2-(2H-1,2,3-benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol |
| SMILES | CC(C)(C)CC(C)(C)C1=CC=C(O)C(=C1)N1N=C2C=CC=CC2=N1 |
Chemical Identifiers
| 3147-75-9 | |
| 323.44 | |
| IYAZLDLPUNDVAG-UHFFFAOYSA-N | |
| 2-(2H-1,2,3-benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol |
| C20H25N3O | |
| MFCD00013338 | |
| 62485 | |
| CC(C)(C)CC(C)(C)C1=CC=C(O)C(=C1)N1N=C2C=CC=CC2=N1 |
Specifications
| 3147-75-9 | |
| White-Yellow | |
| MFCD00013338 | |
| IYAZLDLPUNDVAG-UHFFFAOYSA-N | |
| 2-(2H-1,2,3-benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol | |
| 62485 | |
| ≥98.0% (HPLC,T) | |
| 2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole |
| 104°C | |
| C20H25N3O | |
| 25 g | |
| CC(C)(C)CC(C)(C)C1=CC=C(O)C(=C1)N1N=C2C=CC=CC2=N1 | |
| 323.44 | |
| 323.44 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-5371
TSCA : Yes