missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 104°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 323.44 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole |
Safety and Handling
| EINECSNumber | (5)-5371 |
| TSCA | Yes |