missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Hydroxy-5-methylphenyl)benzotriazole 99.0+%, TCI America™
2-(2-Hydroxy-5-methylphenyl)benzotriazole, >-99.0% (HPLC,T), C13H11N3O, CAS Number-2440-22-4
$115.57 - $419.41
Chemical Identifiers
| CAS | 2440-22-4 |
|---|---|
| Molecular Formula | C13H11N3O |
| Molecular Weight (g/mol) | 225.251 |
| MDL Number | MFCD00022903 |
| InChI Key | MCPKSFINULVDNX-UHFFFAOYSA-N |
| Synonym | drometrizole, benazol p, tinuvin p, 2-2-benzotriazolyl-p-cresol, uv absorber-1, 2-2-hydroxy-5-methylphenyl benzotriazole, 2-2h-benzotriazol-2-yl-p-cresol, 2-2h-benzo d 1,2,3 triazol-2-yl-4-methylphenol, tin p, porex p |
| PubChem CID | 17113 |
| IUPAC Name | 2-(benzotriazol-2-yl)-4-methylphenol |
| SMILES | CC1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2 |
Description
2-(2-Hydroxy-5-methylphenyl)benzotriazole, >-99.0% (HPLC,T), C13H11N3O, CAS Number-2440-22-4
Chemical Identifiers
| 2440-22-4 | |
| 225.251 | |
| MCPKSFINULVDNX-UHFFFAOYSA-N | |
| 17113 | |
| CC1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2 |
| C13H11N3O | |
| MFCD00022903 | |
| drometrizole, benazol p, tinuvin p, 2-2-benzotriazolyl-p-cresol, uv absorber-1, 2-2-hydroxy-5-methylphenyl benzotriazole, 2-2h-benzotriazol-2-yl-p-cresol, 2-2h-benzo d 1,2,3 triazol-2-yl-4-methylphenol, tin p, porex p | |
| 2-(benzotriazol-2-yl)-4-methylphenol |
Specifications
| 2440-22-4 | |
| Yellow | |
| MFCD00022903 | |
| drometrizole, benazol p, tinuvin p, 2-2-benzotriazolyl-p-cresol, uv absorber-1, 2-2-hydroxy-5-methylphenyl benzotriazole, 2-2h-benzotriazol-2-yl-p-cresol, 2-2h-benzo d 1,2,3 triazol-2-yl-4-methylphenol, tin p, porex p | |
| CC1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2 | |
| 225.251 | |
| 225.25 | |
| Crystalline Powder |
| 130°C | |
| C13H11N3O | |
| 25 g | |
| MCPKSFINULVDNX-UHFFFAOYSA-N | |
| 2-(benzotriazol-2-yl)-4-methylphenol | |
| 17113 | |
| ≥99.0% (HPLC,T) | |
| 2-(2-Hydroxy-5-methylphenyl)benzotriazole |
Safety and Handling
EINECSNumber : (5)-0544
RTECSNumber : GO6860000
TSCA : Yes