missing translation for 'onlineSavingsMsg'
Learn More

[2-(2-Furyl)phenyl]methanol, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC46209DE

Quantity 5g

 View more versions of this product

Catalog No. CC46209DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

139697-88-4
174.20
NBLMKRIYULDNBF-UHFFFAOYSA-N
10855985
OCC1=CC=CC=C1C1=CC=CO1
C11H10O2
MFCD06203101
2-2-furyl phenyl methanol, 2-furan-2-yl phenyl methanol, 2-fur-2-yl benzyl alcohol, 2-2-furyl benzenemethanol, 2-fur-2-yl phenyl methanol, benzenemethanol,2-2-furanyl, 2-2-furyl phenyl methan-1-ol
[2-(furan-2-yl)phenyl]methanol

Specifications

[2-(2-Furyl)phenyl]methanol
96.5
97%
5g
2-2-furyl phenyl methanol, 2-furan-2-yl phenyl methanol, 2-fur-2-yl benzyl alcohol, 2-2-furyl benzenemethanol, 2-fur-2-yl phenyl methanol, benzenemethanol,2-2-furanyl, 2-2-furyl phenyl methan-1-ol
OCC1=CC=CC=C1C1=CC=CO1
174.20
174.2
139697-88-4
100.0
C11H10O2
MFCD06203101
NBLMKRIYULDNBF-UHFFFAOYSA-N
[2-(furan-2-yl)phenyl]methanol
10855985
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents