accessibility menu, dialog, popup

UserName

HOO

CAS RN 139697-88-4

IUPAC Name: [2-(furan-2-yl)phenyl]methanol
Synonyms: [2-(furan-2-yl)phenyl]methanol
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: NBLMKRIYULDNBF-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)