missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester, 97%

Catalog No. AC445050010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445050010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445050010 Supplier Thermo Scientific Chemicals Supplier No. 445050010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1073371-99-9 | C14H19BO5 | 278.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073371-99-9
Molecular Formula C14H19BO5
Molecular Weight (g/mol) 278.11
MDL Number MFCD08458200
InChI Key SRYGMLVCVMGUTB-UHFFFAOYSA-N
Synonym methyl 2-hydroxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-hydroxy-4-methoxycarbonylphenylboronic acid, pinacol ester, methyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl salicylate, methyl 2-hydroxy-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
PubChem CID 17998928
IUPAC Name methyl 2-hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES COC(=O)C1=C(O)C=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 85.0°C to 87.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 278.12
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only