missing translation for 'onlineSavingsMsg'
Learn More

(2-[2-(Fmoc-amino)ethoxy]ethoxy)acetic acid, 95%

Catalog No. AAH63641MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63641MD 250 mg
AAH6364103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63641MD Supplier Thermo Scientific Chemicals Supplier No. H63641MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 166108-71-0 | C21H23NO6 | 385.416 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 166108-71-0
Molecular Formula C21H23NO6
Molecular Weight (g/mol) 385.416
MDL Number MFCD01321015
InChI Key XQPYRJIMPDBGRW-UHFFFAOYSA-N
Synonym 2-2-fmoc-amino ethoxy ethoxy acetic acid, fmoc-nh-peg2-ch2cooh, fmoc-aeea, 1-9h-fluoren-9-yl-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, 8-fmoc-amino-3,6-dioxaoctanoic acid, fmoc-adoa, fmoc-aeeac-oh, fmoc-amino-3,6 dioxaoctanoic acid, 8-fmoc-amino-3,6-dioxa-n-octanoic acid
PubChem CID 2756092
IUPAC Name 2-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]acetic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCOCCOCC(=O)O

Specifications

Color White
Linear Formula FmocNHCH2CH2OCH2CH2OCH2CO2H
Quantity 250 mg
Beilstein 9658732
Formula Weight 385.42
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (2-[2-(Fmoc-amino)ethoxy]ethoxy)acetic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only