missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. F10855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10855G 5 g
F108525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10855G Supplier TCI America Supplier No. F10855G

Chemical Identifiers

CAS 876062-39-4
Molecular Formula C12H16BFO2
Molecular Weight (g/mol) 222.07
MDL Number MFCD05663874
InChI Key RZYXBJMVAFMMLQ-UHFFFAOYSA-N
Synonym 1-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-Fluorophenylboronic Acid Pinacol Ester
PubChem CID 12159722
IUPAC Name 2-(2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 222.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane