missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy) Ethyl Acetate, 99%, Spectrum™ Chemical

$390.77 - $15767.70

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
InChI Key FPZWZCWUIYYYBU-UHFFFAOYSA-N
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(C)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18608138
View Documents
Spectrum Chemical Mfg Cor
E10231LT
1 L Amber Glass Bottle
Each for $390.77
Only null left
 
18608139
View Documents
Spectrum Chemical Mfg Cor
E10234LTGL
4 L Amber Glass Bottle
Each for $1,166.80
Only null left
 
18608140
View Documents
Spectrum Chemical Mfg Cor
E102320LTMT
20 L Steel Can
Each for $3,298.97
Only null left
 
18608141
View Documents
Spectrum Chemical Mfg Cor
E1023200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

112-15-2
176.21
2-(2-ethoxyethoxy)ethyl acetate
C8H16O4
FPZWZCWUIYYYBU-UHFFFAOYSA-N
CCOCCOCCOC(C)=O

Specifications

0.0002
1
C8H16O4
1.008 - 1.013
CCOCCOCCOC(C)=O
176.21
Reagent
112-15-2
Amber Glass Bottle
1 L
FPZWZCWUIYYYBU-UHFFFAOYSA-N
2-(2-ethoxyethoxy)ethyl acetate
0.99
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.