missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy) Ethyl Acetate, 99%, Spectrum™ Chemical

Catalog No. 18608138
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608138 1 L Amber Glass Bottle
18608139 4 L Amber Glass Bottle
18608140 20 L Amber Glass Bottle
18608141 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608138 Supplier Spectrum Chemical Mfg Cor Supplier No. E10231LT

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
InChI Key FPZWZCWUIYYYBU-UHFFFAOYSA-N
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(C)=O

Specifications

Acidity 0.0002
CAS Max % 1
Boiling Point 215°C (419°F)
Packaging Amber Glass Bottle
Quantity 1 L
Specific Gravity 1.008 - 1.013
Percent Purity 0.99
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-(2-Ethoxyethoxy) Ethyl Acetate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.