missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dithiodipropionic Acid 98.0+%, TCI America™

Catalog No. D09465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09465G 5 g
D094625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D09465G Supplier TCI America Supplier No. D09465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4775-93-3
Molecular Formula C6H10O4S2
Molecular Weight (g/mol) 210.262
MDL Number MFCD00059649
InChI Key UTDRPUGFHHVEDG-UHFFFAOYSA-N
PubChem CID 521224
IUPAC Name 2-(1-carboxyethyldisulfanyl)propanoic acid
SMILES CC(C(=O)O)SSC(C)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 210.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dithiodipropionic Acid