missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dithiodibenzoic Acid 96.0+%, TCI America™

Catalog No. p-7126727
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D143725G 25 g
D1437100G 100 g
D1437500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D143725G Supplier TCI America Supplier No. D143725G
Only null left

Chemical Identifiers

CAS 119-80-2
Molecular Formula C14H10O4S2
Molecular Weight (g/mol) 306.35
MDL Number MFCD00002465
InChI Key LBEMXJWGHIEXRA-UHFFFAOYSA-N
Synonym 2,2'-dithiodibenzoic acid, 2,2'-dithiosalicylic acid, benzoic acid, 2,2'-dithiobis, 2-2-carboxyphenyl disulfanyl benzoic acid, bis 2-carboxyphenyl disulfide, bis o-carboxylphenyl disulfide, 2,2-dithiosalicylic acid, 2,2'-dithiobisbenzoic acid, unii-58mgb5279d, 2,2'-dithiodi benzoic acid
PubChem CID 8409
IUPAC Name 2-[(2-carboxyphenyl)disulfanyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O

Specifications

Quantity 25 g
Formula Weight 306.35
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dithiodibenzoic Acid
EINECSNumber (3)-3077&(9)-1865
RTECSNumber DG9660000
TSCA Yes