missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dithiodianiline 98.0+%, TCI America™

Catalog No. p-7126646
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D124625G 25 g
D1246500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D124625G Supplier TCI America Supplier No. D124625G
Only null left

Chemical Identifiers

CAS 1141-88-4
Molecular Formula C12H12N2S2
Molecular Weight (g/mol) 248.36
MDL Number MFCD00007703
InChI Key YYYOQURZQWIILK-UHFFFAOYSA-N
Synonym 2,2'-dithiodianiline, benzenamine, 2,2'-dithiobis, intramine, 2,2'-disulfanediyldianiline, 2-aminophenyl disulfide, 2,2'-diaminodiphenyldisulfide, 2,2'-diaminodiphenyl disulfide, 2-2-aminophenyl disulfanyl aniline, aniline, 2,2'-dithiodi, bis o-aminophenyl disulfide
PubChem CID 14358
IUPAC Name 2-[(2-aminophenyl)disulfanyl]aniline
SMILES NC1=CC=CC=C1SSC1=CC=CC=C1N

Specifications

Melting Point 92°C
Quantity 25 g
Formula Weight 248.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dithiodianiline
EINECSNumber (3)-1128
RTECSNumber BX9540000
TSCA Yes