missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Diphenylpropionitrile 98.0+%, TCI America™
Supplier: TCI America D253425G
Specifications
| 2,2-Diphenylpropionitrile | |
| Yellow | |
| C15H13N | |
| 25 g | |
| DPVHBXFSKLKYIQ-UHFFFAOYSA-N | |
| 2,2-diphenylpropanenitrile | |
| 79677 | |
| ≥98.0% (GC) |
| 5558-67-8 | |
| 142°C | |
| MFCD00001846 | |
| 2,2-diphenylpropionitrile, 1,1-diphenylethyl cyanide, 2,2-diphenylpropiononitrile, 2,2-diphenyl propionitrile, .alpha.,.alpha.-diphenylpropionitrile, pubchem12547, acmc-209lnw, 2,2-diphenylpropanitrile, 2,2-diphenyl-propionitrile, 2, 2-diphenyl propionitrile | |
| CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2 | |
| 207.276 | |
| 207.28 | |
| Liquid |
Chemical Identifiers
| 5558-67-8 | |
| 207.276 | |
| DPVHBXFSKLKYIQ-UHFFFAOYSA-N | |
| 79677 | |
| CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2 |
| C15H13N | |
| MFCD00001846 | |
| 2,2-diphenylpropionitrile, 1,1-diphenylethyl cyanide, 2,2-diphenylpropiononitrile, 2,2-diphenyl propionitrile, .alpha.,.alpha.-diphenylpropionitrile, pubchem12547, acmc-209lnw, 2,2-diphenylpropanitrile, 2,2-diphenyl-propionitrile, 2, 2-diphenyl propionitrile | |
| 2,2-diphenylpropanenitrile |
Safety and Handling
TSCA : No