missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylpropionitrile 98.0+%, TCI America™

Catalog No. D25345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25345G 5 g
D253425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25345G Supplier TCI America Supplier No. D25345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5558-67-8
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00001846
InChI Key DPVHBXFSKLKYIQ-UHFFFAOYSA-N
Synonym 2,2-diphenylpropionitrile, 1,1-diphenylethyl cyanide, 2,2-diphenylpropiononitrile, 2,2-diphenyl propionitrile, .alpha.,.alpha.-diphenylpropionitrile, pubchem12547, acmc-209lnw, 2,2-diphenylpropanitrile, 2,2-diphenyl-propionitrile, 2, 2-diphenyl propionitrile
PubChem CID 79677
IUPAC Name 2,2-diphenylpropanenitrile
SMILES CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
Formula Weight 207.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Diphenylpropionitrile