missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylpropionitrile, 97%

Catalog No. AAA1799118
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1799118 50 g
AAA1799106 5 g
AAA1799109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1799118 Supplier Thermo Scientific Chemicals Supplier No. A1799118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5558-67-8 | C15H13N | 207.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5558-67-8
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00001846
InChI Key DPVHBXFSKLKYIQ-UHFFFAOYSA-N
PubChem CID 79677
IUPAC Name 2,2-diphenylpropanenitrile
SMILES CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Density 1.108
Boiling Point 144°C to 145°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.572
Quantity 50 g
UN Number UN3276
Beilstein 1955952
Synonym 2,2-diphenylpropionitrile, 1,1-diphenylethyl cyanide, 2,2-diphenylpropiononitrile, 2,2-diphenyl propionitrile, .alpha.,.alpha.-diphenylpropionitrile, pubchem12547, acmc-209lnw, 2,2-diphenylpropanitrile, 2,2-diphenyl-propionitrile, 2, 2-diphenyl propionitrile
Formula Weight 207.28
Percent Purity 97%
Chemical Name or Material 2,2-Diphenylpropionitrile
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 226-925-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only