missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylacetamide, 98%

Catalog No. aal0848514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0848514 25 g
AAL0848506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0848514 Supplier Thermo Scientific Chemicals Supplier No. L0848514

CAS: 4695-13-0 | C14H13NO | 211.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4695-13-0
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.264
MDL Number MFCD00025496
InChI Key ZXQVXEAZKZFEEP-UHFFFAOYSA-N
Synonym diphenylacetic acid amide, acetamide, 2,2-diphenyl, alpha-phenylbenzeneacetamide, alpha,alpha-diphenylacetamide, unii-yzx41252dl, 2,2-diphenylethanamide, benzeneacetamide, .alpha.-phenyl, .alpha.,.alpha.-diphenylacetamide, acmc-20aojh, 2,2-diphenylacetamid
PubChem CID 78420
IUPAC Name 2,2-diphenylacetamide
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)N

Specifications

Melting Point 164°C to 167°C
Quantity 25 g
Beilstein 1871583
Merck Index 14,3316
Formula Weight 211.26
Percent Purity 98%
Chemical Name or Material 2,2-Diphenylacetamide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber AB8135000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only