missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylacetamide, 98%

CAS: 4695-13-0 | C14H13NO | 211.264 g/mol

Supplier:  Thermo Scientific Chemicals L0848506

 View more versions of this product

Catalog No. AAL0848506

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,2-Diphenylacetamide
164°C to 167°C
MFCD00025496
1871583
diphenylacetic acid amide, acetamide, 2,2-diphenyl, alpha-phenylbenzeneacetamide, alpha,alpha-diphenylacetamide, unii-yzx41252dl, 2,2-diphenylethanamide, benzeneacetamide, .alpha.-phenyl, .alpha.,.alpha.-diphenylacetamide, acmc-20aojh, 2,2-diphenylacetamid
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)N
211.264
211.26
4695-13-0
C14H13NO
5 g
14,3316
ZXQVXEAZKZFEEP-UHFFFAOYSA-N
2,2-diphenylacetamide
78420
98%

Chemical Identifiers

4695-13-0
211.264
ZXQVXEAZKZFEEP-UHFFFAOYSA-N
78420
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)N
C14H13NO
MFCD00025496
diphenylacetic acid amide, acetamide, 2,2-diphenyl, alpha-phenylbenzeneacetamide, alpha,alpha-diphenylacetamide, unii-yzx41252dl, 2,2-diphenylethanamide, benzeneacetamide, .alpha.-phenyl, .alpha.,.alpha.-diphenylacetamide, acmc-20aojh, 2,2-diphenylacetamid
2,2-diphenylacetamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

RTECSNumber : AB8135000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only