missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dinaphthylamine 98.0+%, TCI America™
$152.86 - $456.70
Chemical Identifiers
| CAS | 532-18-3 |
|---|---|
| Molecular Formula | C20H15N |
| Molecular Weight (g/mol) | 269.347 |
| MDL Number | MFCD03093264 |
| InChI Key | SBMXAWJSNIAHFR-UHFFFAOYSA-N |
| PubChem CID | 68283 |
| IUPAC Name | N-naphthalen-2-ylnaphthalen-2-amine |
| SMILES | C1=CC=C2C=C(C=CC2=C1)NC3=CC4=CC=CC=C4C=C3 |
Chemical Identifiers
| 532-18-3 | |
| 269.347 | |
| SBMXAWJSNIAHFR-UHFFFAOYSA-N | |
| N-naphthalen-2-ylnaphthalen-2-amine |
| C20H15N | |
| MFCD03093264 | |
| 68283 | |
| C1=CC=C2C=C(C=CC2=C1)NC3=CC4=CC=CC=C4C=C3 |
Specifications
| 532-18-3 | |
| 471°C | |
| MFCD03093264 | |
| 2,2′C-Iminodinaphthalene | |
| C1=CC=C2C=C(C=CC2=C1)NC3=CC4=CC=CC=C4C=C3 | |
| 269.347 | |
| 269.35 | |
| Crystalline Powder |
| 174°C | |
| C20H15N | |
| 1 g | |
| SBMXAWJSNIAHFR-UHFFFAOYSA-N | |
| N-naphthalen-2-ylnaphthalen-2-amine | |
| 68283 | |
| ≥98.0% (GC) | |
| 2,2′-Dinaphthylamine |
Safety and Handling
TSCA : No