missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dinaphthylamine 98.0+%, TCI America™
Supplier: TCI America D29875G
Specifications
| 2,2′-Dinaphthylamine | |
| 174°C | |
| C20H15N | |
| 5 g | |
| SBMXAWJSNIAHFR-UHFFFAOYSA-N | |
| N-naphthalen-2-ylnaphthalen-2-amine | |
| 68283 | |
| ≥98.0% (GC) |
| 532-18-3 | |
| 471°C | |
| MFCD03093264 | |
| 2,2′-Iminodinaphthalene | |
| C1=CC=C2C=C(C=CC2=C1)NC3=CC4=CC=CC=C4C=C3 | |
| 269.347 | |
| 269.35 | |
| Crystalline Powder |
Chemical Identifiers
| 532-18-3 | |
| 269.347 | |
| SBMXAWJSNIAHFR-UHFFFAOYSA-N | |
| N-naphthalen-2-ylnaphthalen-2-amine |
| C20H15N | |
| MFCD03093264 | |
| 68283 | |
| C1=CC=C2C=C(C=CC2=C1)NC3=CC4=CC=CC=C4C=C3 |
Safety and Handling
TSCA : No