Learn More
2,2-Dimethylhexanoic acid, 94%, Thermo Scientific Chemicals
$137.37
Chemical Identifiers
| CAS | 813-72-9 |
|---|---|
| Molecular Formula | C8H16O2 |
| Molecular Weight (g/mol) | 144.21 |
| MDL Number | MFCD00965097 |
| InChI Key | YTTWDTVYXAEAJA-UHFFFAOYSA-N |
| Synonym | 2,2-dimethyl-hexanoic acid, hexanoic acid,2,2-dimethyl, 2,2-dimethylhexanoicacid, dimethylhexanoic acid, dimethyl-hexanoic acid, acmc-209plb, 2,2-dimethyl hexanoic acid, #, hexanoic acid, 2,2-dimethyl, yttwdtvyxaeaja-uhfffaoysa |
| PubChem CID | 163217 |
| IUPAC Name | 2,2-dimethylhexanoic acid |
| SMILES | CCCCC(C)(C)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL1466403
|
N/A | 1 g |
Each for $137.37
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C8H16O2 | |
| MFCD00965097 | |
| 2,2-dimethyl-hexanoic acid, hexanoic acid,2,2-dimethyl, 2,2-dimethylhexanoicacid, dimethylhexanoic acid, dimethyl-hexanoic acid, acmc-209plb, 2,2-dimethyl hexanoic acid, #, hexanoic acid, 2,2-dimethyl, yttwdtvyxaeaja-uhfffaoysa | |
| 2,2-dimethylhexanoic acid |
Specifications
| 813-72-9 | |
| 216°C to 220°C | |
| 1.427 | |
| UN3265 | |
| 2,2-dimethyl-hexanoic acid, hexanoic acid,2,2-dimethyl, 2,2-dimethylhexanoicacid, dimethylhexanoic acid, dimethyl-hexanoic acid, acmc-209plb, 2,2-dimethyl hexanoic acid, #, hexanoic acid, 2,2-dimethyl, yttwdtvyxaeaja-uhfffaoysa | |
| CCCCC(C)(C)C(O)=O | |
| 144.21 | |
| 144.22 | |
| 2,2-Dimethylhexanoic acid |
| 0.913 | |
| C8H16O2 | |
| MFCD00965097 | |
| 1749961 | |
| YTTWDTVYXAEAJA-UHFFFAOYSA-N | |
| 2,2-dimethylhexanoic acid | |
| 163217 | |
| 94% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only