missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylglutaric Anhydride 98.0+%, TCI America™

Catalog No. p-7126677
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13255G 5 g
D132525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13255G Supplier TCI America Supplier No. D13255G

Chemical Identifiers

CAS 2938-48-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00006682
InChI Key PAVNZLVXYJDFNR-UHFFFAOYSA-N
Synonym 2,2-dimethylglutaric anhydride, 2h-pyran-2,6 3h-dione, dihydro-3,3-dimethyl, 3,3-dimethyldihydro-2h-pyran-2,6 3h-dione, dihydro-3,3-dimethyl-2h-pyran-2,6 3h-dione, .alpha.,.alpha.-dimethyl glutaric anhydride, 3,3-dimethyltetrahydro-2h-pyran-2,6-dione, acmc-209h8l, 2,2-dimethylglutaricanhydride, 2,2 dimethylglutaric anhydride, 2.2-dimethylglutaric anhydride
PubChem CID 76247
IUPAC Name 3,3-dimethyloxane-2,6-dione
SMILES CC1(C)CCC(=O)OC1=O

Specifications

Melting Point 38°C
Color White
Boiling Point 270°C
Quantity 5 g
Formula Weight 142.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dimethylglutaric Anhydride