accessibility menu, dialog, popup

UserName

CH3H3COOO

CAS RN 2938-48-9

IUPAC Name: 3,3-dimethyloxane-2,6-dione
Synonyms: 3,3-dimethyloxane-2,6-dione
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: PAVNZLVXYJDFNR-UHFFFAOYSA-N
SMILES: CC1(C)CCC(=O)OC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results