missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylglutaric Acid 98.0+%, TCI America™

Catalog No. D178625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D178625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D178625G Supplier TCI America Supplier No. D178625G

Chemical Identifiers

CAS 681-57-2
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00004197
InChI Key BTUDGPVTCYNYLK-UHFFFAOYSA-N
Synonym 2,2-dimethylglutaric acid, 2,2-dimethylglutaricacid, unii-s46amq2g6c, pentanedioic acid, dimethyl, s46amq2g6c, glutaric acid, 2,2-dimethyl, .alpha.,.alpha.-dimethylglutaric acid, pentanedioic acid, 2,2-dimethyl, dimethylglutaric acid, dsstox_cid_1585
PubChem CID 12681
ChEBI CHEBI:68459
IUPAC Name 2,2-dimethylpentanedioic acid
SMILES CC(C)(CCC(=O)O)C(=O)O

Specifications

Melting Point 86°C
Color White
Boiling Point 200°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dimethylglutaric Acid
EINECSNumber (2)-2561
TSCA No