accessibility menu, dialog, popup

UserName

CAS RN 681-57-2

H3CCH3HOOOHO

CAS RN 681-57-2

IUPAC Name: 2,2-dimethylpentanedioic acid
Synonyms: glutaric acid, 2,2-dimethyl-
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: BTUDGPVTCYNYLK-UHFFFAOYSA-N
SMILES: CC(C)(CCC(O)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results