missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylcyclopropyl Cyanide 97.0+%, TCI America™

Catalog No. C15241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15241G 1 g
C15245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15241G Supplier TCI America Supplier No. C15241G

Chemical Identifiers

CAS 5722-11-2
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD01321153
InChI Key CZPCJHBAFHCDRE-UHFFFAOYNA-N
Synonym 1-Cyano-2,2-dimethylcyclopropane
PubChem CID 260707
IUPAC Name 2,2-dimethylcyclopropane-1-carbonitrile
SMILES CC1(CC1C#N)C

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Formula Weight 95.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylcyclopropyl Cyanide