accessibility menu, dialog, popup

UserName

CH3H3CN

CAS RN 5722-11-2

IUPAC Name: 2,2-dimethylcyclopropane-1-carbonitrile
Synonyms: 2,2-dimethylcyclopropane-1-carbonitrile
Molecular Weight (g/mol): 95.15
Molecular Formula: C6H9N
InChi Key: CZPCJHBAFHCDRE-UHFFFAOYNA-N
SMILES: CC1(C)CC1C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results