missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[[2-(Dimethylamino)ethyl]methylamino]ethanol 97.0+%, TCI America™
$76.43 - $197.59
Chemical Identifiers
| CAS | 2212-32-0 |
|---|---|
| Molecular Formula | C7H18N2O |
| Molecular Weight (g/mol) | 146.23 |
| MDL Number | MFCD00010387 |
| InChI Key | LSYBWANTZYUTGJ-UHFFFAOYSA-N |
| PubChem CID | 75171 |
| IUPAC Name | 2-{[2-(dimethylamino)ethyl](methyl)amino}ethan-1-ol |
| SMILES | CN(C)CCN(C)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D468025ML
|
TCI America
D468025ML |
25 mL |
Each for $76.43
|
|
|||||
|
D4680100ML
|
TCI America
D4680100ML |
100 mL |
Each for $197.59
|
|
|||||
Chemical Identifiers
| 2212-32-0 | |
| 146.23 | |
| LSYBWANTZYUTGJ-UHFFFAOYSA-N | |
| 2-{[2-(dimethylamino)ethyl](methyl)amino}ethan-1-ol |
| C7H18N2O | |
| MFCD00010387 | |
| 75171 | |
| CN(C)CCN(C)CCO |
Specifications
| 2212-32-0 | |
| 88°C | |
| MFCD00010387 | |
| N′C-(2-Hydroxyethyl)-N,N,N′C-trimethylethylenediamine | |
| CN(C)CCN(C)CCO | |
| 146.23 | |
| 146.23 | |
| Liquid |
| Yellow | |
| C7H18N2O | |
| 25 mL | |
| LSYBWANTZYUTGJ-UHFFFAOYSA-N | |
| 2-{[2-(dimethylamino)ethyl](methyl)amino}ethan-1-ol | |
| 75171 | |
| ≥97.0% (GC,T) | |
| 2-[[2-(Dimethylamino)ethyl]methylamino]ethanol |
Safety and Handling
EINECSNumber : (2)-3206
TSCA : Yes