accessibility menu, dialog, popup

UserName

CAS RN 2212-32-0

H3CNCH3NCH3OH

CAS RN 2212-32-0

IUPAC Name: 2-{[2-(dimethylamino)ethyl](methyl)amino}ethan-1-ol
Synonyms: 2-{[2-(dimethylamino)ethyl](methyl)amino}ethanol
Molecular Weight (g/mol): 146.23
Molecular Formula: C7H18N2O
InChi Key: LSYBWANTZYUTGJ-UHFFFAOYSA-N
SMILES: CN(C)CCN(C)CCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results