missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[[2-(Dimethylamino)ethyl]methylamino]ethanol 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 88°C |
| Quantity | 25 mL |
| Synonym | N′C-(2-Hydroxyethyl)-N,N,N′C-trimethylethylenediamine |
| Formula Weight | 146.23 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-[[2-(Dimethylamino)ethyl]methylamino]ethanol |
Safety and Handling
| EINECSNumber | (2)-3206 |
| TSCA | Yes |